missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DynaMag™-PCR Magnet

Catalog No. 501148769
Encompass
Change view
Click to view available options
Quantity:
1 each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501148769 1 each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 501148769 Supplier Invitrogen™ Supplier No. 492025

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The MAGnify™ ChIP system utilizes a novel magnet that is compatible with 0.2mL PCR strip tubes.

The MAGnify™ ChIP system utilizes a novel magnet that is compatible with 0.2mL PCR strip tubes. This magnet is critical for enabling the use of reduced starting cell numbers and volumes for immunoprecipitation and washing steps. This novel magnetic separator allows you to achieve results quickly and easily while offering the freedom to simultaneously perform multiple ChIP assays in PCR tubes with a multi-channel pipettor.

Specifications

Content And Storage Store magnet at room temperature.
For Use With (Equipment) DynaMag™
Product Line DynaMag
Quantity 1 each
Shipping Condition Room Temperature
What are the Gauss levels for the DynaMag-2 and DynaMag-Spin Magnets?

The DynaMag-2 Magnet is 3,500-3,700 minimum Gauss level and the DynaMag-Spin is 3,000-3,500 minimum Gauss level.

What is the standard conversion of Gauss to Tesla units?

1 Tesla is equivalent to 10,000 Gauss. .

How can I clean my DynaMag magnet?

Spray and/or wipe the DynaMag magnet unit with one of the following cleaning agents:

- 70% (v/v) isopropyl alcohol
- 1% (v/v) sodium hypochlorite solution (bleach)
- 0.1 N HCl solution

Do not submerge the magnet unit in aqueous solutions and avoid prolonged exposure to water or aqueous solutions. Clean with a damp cloth and mild detergent when exposed to harsh solvents. Do not autoclave the magnet unit as heating the magnet to above 70 degrees C can destabilize the magnetic material, resulting in loss of magnetic strength.

What is the difference between the DynaMag-2 Magnet (Cat. No. 12321D) and DynaMag-Spin Magnet (Cat. No. 12320D)?

The DynaMag-2 Magnet can hold 16 standard 1.5 mL or 2 mL microcentrifuge tubes, while the DynaMag-Spin Magnet holds up to 6 standard 1.5 mL microcentrifuge tubes.

How can I clean my DynaMag magnet?

Spray and/or wipe the DynaMag magnet unit with one of the following cleaning agents:

- 70% (v/v) isopropyl alcohol
- 1% (v/v) sodium hypochlorite solution (bleach)
- 0.1 N HCl solution

Do not submerge the magnet unit in aqueous solutions and avoid prolonged exposure to water or aqueous solutions. Clean with a damp cloth and mild detergent when exposed to harsh solvents. Do not autoclave the magnet unit as heating the magnet to above 70 degrees C can destabilize the magnetic material, resulting in loss of magnetic strength.

For Research Use Only. Not for use in diagnostic procedures.