missing translation for 'onlineSavingsMsg'
Learn More

Dynalon™ Bio-Bin™ Waste Disposal Containers

A new cost effective and convenient alternative to rigid plastic waste collection and disposal containers

$446.38 - $1,166.48

Specifications

Certifications/Compliance UN 3291
For Use With (Application) Ideal solution for biohazard or non-hazardous waste
Product Type Waste Container
View More Specs

Products 3
Catalog Number Mfr. No. Capacity (English) Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (English) Price Quantity & Availability  
12009801
Accessories View Documents Promotion Details
N/A 1.5 gal.
Case of 40 Each for $1,166.48
Only null left
 
12009802
View Documents Promotion Details
Dynalon™
7973030030
7.9 gal.
Case of 10 Each for $446.38
Only null left
 
12009800
View Documents Promotion Details
N/A 0.66 gal.
Case of 50 Each for $446.38
Only null left
 
Description

Description

  • Space efficient, light weight fold-flat design
  • Built-in lid and handles
  • Waterproof construction
  • Temporary and permanent seals
  • Built-in super absorbent Bio-matt™ to soak up any excess liquid
  • Autoclavable (use with #202065 Autoclave Sterility Tape)
  • Supplied with 2 x 2 in. Biohazard Waste Labels
  • Stackable post use (UN 3291 Approved)

Order Info

Accessories available. (See Cat. No. 12-009-803)

Specifications

Specifications

UN 3291
Ideal solution for biohazard or non-hazardous waste
Waste Container
Videos
SDS
Documents

Documents