Learn More
Dynalon™ Bio-Bin™ Waste Disposal Containers
A new cost effective and convenient alternative to rigid plastic waste collection and disposal containers
Description
- Space efficient, light weight fold-flat design
- Built-in lid and handles
- Waterproof construction
- Temporary and permanent seals
- Built-in super absorbent Bio-matt™ to soak up any excess liquid
- Autoclavable (use with #202065 Autoclave Sterility Tape)
- Supplied with 2 x 2 in. Biohazard Waste Labels
- Stackable post use (UN 3291 Approved)
Order Info
Accessories available. (See Cat. No. 12-009-803)
Chemical Identifiers
| UN 3291 | |
| Ideal solution for biohazard or non-hazardous waste | |
| Waste Container |
Specifications
| 2.5 L | |
| 0.66 gal. | |
| 24.38 cm | |
| 10.9 cm | |
| Ideal solution for biohazard or non-hazardous waste | |
| 9 in. | |
| Waste Container |
| UN 3291 | |
| 9 x 4.3 x 9.6 in. (22.86 x 10.9 x 24.38 cm) | |
| 22.86 cm | |
| 2 x 2 in. biohazard waste labels, 1.5mL thick polyethylene bag | |
| 4.3 in. | |
| 9.6 in. |