Learn More
Dynalon™ Automatic Glass Titrating Burets
Supplier:  Dynalon™ 8969541025
 View more versions of this product
Catalog No. 02550103
Only null left
Description
Description
Simple and accurate to use. Squeeze the flexible LDPE bottle until the titrate solution flows through the tubing into the glass buret to just above the “0” graduation mark. Product is designed to self zero and does not overflow.
Specifications
Chemical Identifiers
Borosilicate Glass
Blue
Class B
Stand, Blue Ring Graduations, Schellbach Stripe
LPDE
DIN ISO 385
Specifications
25 mL
1L LDPE bottle
0.1 mL
61 cm
Blue
±0.1 mL
Class B
24.1 in.
Borosilicate Glass
Electroplating Industry, Field Analyses and Numerous Other Applications, Universal Aid in the Laboratory, Water Treatment Plants
Stand, Blue Ring Graduations, Schellbach Stripe
LPDE
Pellet Pattern Automatic Buret
DIN ISO 385
Product Suggestions
Product Suggestions