missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ DyLight™ 550 Maleimide

Catalog No. PI62290
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI62290 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI62290 Supplier Thermo Scientific™ Supplier No. 62290

Label antibodies or other proteins as probes for fluorescence imaging assays with the full spectral range of maleimide-activated DyLight Dyes.

Thermo Scientific™ Sulfhydryl-Reactive DyLight Dyes are maleimide-activated DyLight Fluors for fluorescent labeling of antibodies and other proteins to be used as probes for cellular imaging and other fluorescence detection methods.

The Thermo Scientific™ DyLight™ Dyes have absorption spectra ranging from 350 to 770nm. These reagents fluoresce over a broad pH range, are more intense than Alexa Fluor™ or Cy™ Dyes in many applications and match the output wavelengths of common fluorescence instrumentation. Additionally, the water solubility of the DyLight Reagents allows a high fluor-to-protein ratio without precipitation during conjugation.
The sulfhydryl-reactive dyes contain maleimide groups that react predominantly with free –SH groups at pH 6.5-7.5, forming a stable thioether bond. At pH 7, the maleimide group is ∼1000 times more reactive toward a free sulfhydryl than to an amine.1At pH values > 7.5, reactivity toward primary amines increases and hydrolysis of the maleimide group can occur.

Highlights:
  • Specific — maleimide-activated dye labels proteins and other molecules at reduced sulfhydryls (-SH)
  • Efficient labeling methods — well-characterized chemistry and optimized protocols provide for reliable, high-quality labeling
  • Optimized antibody labeling procedure —complete protocol for IgG reduction and labeling and calculating the labeling efficiency

Applications:
  • Antibody labeling for immunofluorescence applications, including immunocytochemistry (ICC), immunohistochemistry (IHC), Western blotting and ELISA assay
  • Target macromolecule labeling for in vitro and in vivo fluorescent detection strategies

Specifications

Description DyLight 550 Maleimide
Quantity 1 mg
Product Type Dye
Solubility DMF (Dimethylformamide)
Content And Storage Store desiccated at -20°C. Shipped at ambient temperature.
Chemical Reactivity Thiol
Reactive Moiety Maleimide
Label or Dye DyLight™ 550
Label Type DyLight Fluors
Product Line DyLight
Emission 576
Excitation 562
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.