missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ DyLight™ 515-LS NHS Ester

Catalog No. PI82491
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI82491 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI82491 Supplier Thermo Scientific™ Supplier No. 82491

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Label antibodies or other proteins for FRET and multiplex fluorescence assays using fluors with large Stokes shifts (488nm excitation; yellow to far-red emission).

Thermo Scientific™ DyLight Long Stokes Shift Specialty Dyes offer a broad spectrum of fluorescence emission characteristics from green to far red with a single excitation peak that matches the 488 laser line and extraordinarily large Stokes shifts that inhibit quenching.

DyLight Long Stokes Shift Specialty Dyes are a family of labeling agents that provide bright fluorescent detection for multiplexed imaging. The dyes share a common excitation range, while their emission properties extend from the green to the far red wavelengths. The dyes are based on a core coumarin structure that is modified to emit at different wavelengths. Each dye contains an amine-reactive NHS ester for rapid modification of antibodies, proteins, peptides or other biomolecules through amide bond formation. DyLight Long Stokes Shift Dyes can be used individually or combined with DyLight 488 for multiplexed analysis. The excitation and emission peaks are sufficiently separated so as to minimize dye-dye quenching in assays.

Highlights:

  • Single excitation wavelength–DyLight Long Stokes Shift Dyes all can be excited together at 488nm using an Argon laser

  • Image multiple targets simultaneously–the four Long Stokes Shift Dyes can be used with DyLight 488 for multiplex fluorescence applications

  • NHS ester reactive group–allows immediate labeling of antibodies, proteins, peptides and other amine-containing molecules through amide bond formation

  • Multiple solubility options –choose from hydrophilic to hydrophobic dyes to optimize the right dye label for the best performance in an application

  • Large Stokes shift inhibits quenching–minimal overlap between excitation and emission peaks inhibits dye-dye quenching effects

  • Use in FRET-based assays– the extremely broad separation between excitation and emission peaks allow for use of the Long Stokes Shift Dyes in energy transfer applications where the acceptor dye is spectrally far removed from the excitation wavelength

Criteria to consider when choosing a DyLight Long Stokes Shift Specialty Dye:

  • Excitation and emission wavelengths–all dyes share a common excitation wavelength range with options for emission from the green to the far red wavelengths

  • Water solubility–DyLight Long Stokes Shift Dyes can be initially solubilized in methanol, ethanol, DMF or DMSO with transfer into an aqueous buffer medium for labeling biomolecules

Recommended for:

Quenchers and FRET pairs; Conjugation to silica particles; STED microscopy; Multicolor microscopy; 4Pi microscopy; Dual-color 3D nanoscopy; Dynamic light scattering; Graded stokes shift; Duplex enhanced RT-PCR; High-resolution optical microscopy

Specifications

Description DyLight 515-LS NHS Ester
Quantity 1 mg
Product Type Dye
Solubility DMF (Dimethylformamide)
Content And Storage Store at -20°C. Shipped at ambient temperature.
Chemical Reactivity Amine
Reactive Moiety Active Ester, Succinimidyl Ester, NHS Ester
Label or Dye DyLight™ Fluor (Specialty)
Label Type DyLight Fluors
Product Line DyLight
Emission 648
Excitation 519
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.