missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ DyLight™ 488, Free Acid

Catalog No. 62280
Encompass
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
62280 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 62280 Supplier Thermo Scientific™ Supplier No. 62280

Use these non-activated, carboxylic acid forms of DyLight Dyes as controls and calibration standards in fluorescence imaging applications.

Thermo Scientific™ DyLight Fluor Free Acids are non-activated forms of selected DyLight Fluorescent Dyes for use as controls and calibration factors in fluorecence imaging applications.

Thermo Scientific™ DyLight Fluor Free Acids are non-activated forms of selected DyLight Fluorescent Dyes for use as controls and calibration factors in fluorecence imaging applications. The non-activated dyes contain the native carboxyl group of the standard dye molecule.

General DyLight Fluorophore Highlights:

  • Bright fluorescence — intense emission provides superior sensitivity and requires less conjugate
  • Narrow emission spectra — negligible bleed-through between fluorophore channels enables multi-color detection
  • Excellent photostability — exceptional resistance to photobleaching enables fluorescence imaging under the most demanding conditions (e.g., structured illumination, 4Pi microscopy)
  • Buffer stability — conjugated fluorophores are completely stable at pH 4-9
  • Instrument compatibility — excitation and emission spectra correspond with filter sets and laser settings of all popular fluorescence instrumentation

Specifications

Description DyLight™ 488, Free Acid, 1mg, Solid, DyLight™ 488 Dye, 493/518nm Excitation/Emission Wavelength, <4°C Storage Temperature, Used for Protein Biology, Protein Labeling and Crosslinking, Protein Labeling
Quantity 1 mg
Product Type Free Acid
Solubility DMF (Dimethylformamide)
Content And Storage Store at < 4°C.
Reactive Moiety Carboxylic Acid
Label or Dye DyLight™ 488
Label Type DyLight Fluors
Product Line DyLight
Emission 518
Excitation 493
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.