missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ DyLight™ Antibody Labeling Kits

Catalog No. PI53020
Change view
Click to view available options
Excitation/Emission:
346/442
400/420
490/525
562/576
590/617
638/658
652/672
679/702
749/775
784/814
Label or Dye:
DyLight 350
DyLight 405
DyLight 488
DyLight 550
DyLight 594
DyLight 633
DyLight 650
DyLight 680
DyLight 755
DyLight 800
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Excitation/Emission Label or Dye
PI53020 400/420 DyLight 405
PI53024 490/525 DyLight 488
PI53044 590/617 DyLight 594
PI53046 638/658 DyLight 633
PI53056 679/702 DyLight 680
PI53062 784/814 DyLight 800
PI62275 346/442 DyLight 350
PI84530 562/576 DyLight 550
PI84535 652/672 DyLight 650
PI84538 749/775 DyLight 755
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. PI53020 Supplier Thermo Scientific™ Supplier No. 53020

Covalently label antibodies or proteins with DyLight dyes for use in immunofluorescence, flow cytometry, western blot analysis, and high content analysis.

Invitrogen Dylight Antibody Labeling kits provide everything needed to label 1 mg of antibody or protein with amine-reactive Dylight dye and subsequent free dye removal. Choose from a variety of Dylight fluorophores across the spectrum to covalently label your antibody or protein. Labeled antibodies are typically ready to use in 90 minutes with minimal hands-on time. The kit contains all necessary components to perform three separate labeling reactions using 1 mg of IgG or similar quantities of other proteins.

Invitrogen DyLight Antibody Labeling kits enable the creation of fluorescent antibodies that are covalently linked to different DyLight fluorophores. The fluorescently labeled antibodies can be used in a wide variety of laboratory applications, including immunofluorescence microscopy, IHC, western blotting, ELISA, flow cytometry, or in vitro and in vivo fluorescent detection strategies.

DyLight Antibody Labeling kits contain an NHS ester-activated derivative of the high-performance DyLight dye that can be used to fluorescently label antibodies and other proteins for cellular imaging and other fluorescence detection methods. The DyLight dye is an amine-reactive dye and is activated with an N-hydroxysuccinimide (NHS) ester moiety to react with exposed N-terminal α-amino groups or the ε-amino groups of lysine residues to form stable amide bonds.

DyLight Antibody Labeling kits contain all necessary components to perform three separate labeling reactions and free dye removal using 1 mg of IgG or similar quantities of other proteins.

DyLight dyes exhibit higher fluorescence intensity and photostability than CyDye and LI-COR™ dyes in many applications and remain highly fluorescent over a broad pH range (pH 4–9). Additionally, the water solubility of the DyLight fluorescent dyes allows a high dye-to-protein ratio to be achieved without causing precipitation of conjugates. Finally, the high solubility of these fluorophores permits protein solutions to be added directly to specific amounts of the labeling reagent.

Other advantages of the DyLight Antibody Labeling kits:
· High performance—high fluorescence intensity across a broad pH range
· Multiple colors—10 different dyes across the visible spectrum and extending into the infrared
· Specificity—NHS ester-activated dye labels proteins and other molecules at primary amines (-NH2)
· Optimized procedure—following the standard protocol results in antibodies with excellent dye:protein ratios and recovery rates for optimum activity and fluorescence labeling

We also offer DyLight Microscale Antibody Labeling kits for fast and efficient fluorescent labeling of antibodies for use in fluorescence methods. The DyLight Microscale Antibody Labeling kits contain all necessary components to perform five separate labeling reactions using 100 μg of IgG or similar quantities of other proteins. Both the standard and microscale kit sizes include the amine-reactive DyLight fluorophore in convenient single-use vials as well as purification resin and spin columns for the preparation of ready-to-use conjugate.

Specifications

Kit Contents Contains 3 vials DyLight NHS Ester, 1 mL Tube Borate Buffer, 5 mL Purification Resin, 6 Spin Columns, and 12 Microcentrifuge Collection Tubes
Product Type Labeling Kit
Quantity 3 reactions
Content And Storage Store the DyLight NHS Ester at -20°C. Store all other kit components at 4°C.
Excitation/Emission 400/420
Labeling Scale 0.5 to 1 mg
Product Line DyLight
Chemical Reactivity Amine
Reactive Moiety NHS
Label or Dye DyLight 405
Color Blue
Labeling Target Antibodies, Proteins
Label Type DyLight™
Show More Show Less