missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ DyLight™ 800 NHS Ester

Catalog No. PI46421
Change view
Click to view available options
Quantity:
5 x 50 μg
1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI46421 1 mg
PI46422 5 x 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PI46421 Supplier Thermo Scientific™ Supplier No. 46421

Label antibodies or other proteins as probes for fluorescence imaging assays with the full spectral range of NHS ester-activated DyLight Dyes.

Thermo Scientific™ Amine-Reactive DyLight Dyes are NHS ester-activated DyLight Fluors for fluorescent labeling of antibodies and other proteins to be used as molecular probes for cellular imaging and other fluorescence detection methods.

The DyLight Dyes exhibit higher fluorescence intensity and photostability than Alexa™, CyDye™ and LI-COR™ Dyes in many applications and remain highly fluorescent over a broad pH range (pH 4-9). Additionally, the water solubility of the DyLight Dyes allows a high dye-to-protein ratio without precipitation during conjugation.

Highlights:

  • Brightness: high-fluorescence intensity provides outstanding sensitivity and requires less conjugate

  • Photostability: exceptional resistance to photobleaching enables fluorescence imaging under the most demanding conditions (e.g., structured illumination, 4pi microscopy)

  • Instrument compatibility: emission spectra match the principal output wavelengths of common fluorescence instrumentation

Specifications

Quantity 1 mg
Product Type 800 NHS Ester
Solubility DMF (Dimethylformamide)
Content And Storage Store desiccated at -20°C.
Chemical Reactivity Amine
Reactive Moiety Active Ester, Succinimidyl Ester, NHS Ester
Label or Dye DyLight™ 800
Label Type DyLight Fluors
Product Line DyLight
Emission 794
Excitation 777
Labeling Target Antibody, Protein
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.