missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ DyLight™ 800-4xPEG NHS Ester

Catalog No. PI46604
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI46604 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI46604 Supplier Thermo Scientific™ Supplier No. LSG46604
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Fluorescently label antibodies and other proteins for near-IR detection with this derivative of our high-performance DyLight 800 Dye.

Fluorescently label antibodies and other proteins for near-IR detection with Thermo Scientific™ DyLight™ 800-4xPEG Amine-Reactive Dye, a derivative of our high-performance DyLight 800 Dye.

Thermo Scientific DyLight 800-4xPEG Amine-Reactive Dye is a derivative of our high-performance DyLight 800 Dye that can be used to fluorescently label antibodies and other proteins for near-IR detection.

The DyLight 800-4xPEG dye contains 4 polyethylene glycol (PEG) chains that are non-cytotoxic, enhance fluorescence and reduce nonspecific binding of conjugates made with them. Conjugates made with DyLight 800-4xPEG Dye can be used as molecular probes for cellular imaging and other fluorescence detection methods. The NIR fluorescence properties of DyLight 800-4xPEG Dye make it especially useful in a variety of biological, chemical, and pharmaceutical applications, including in vivo imaging. The PEG chains also improve solubility of the dyes and labeled molecules in aqueous solution, aid in cell permeability and improve tissue retention.

Highlights:
  • High fluorescence intensity – fluorescence comparable to Alexa Fluor™ 800 and IRDye™ 800
  • PEGylated – improves solubility in aqueous solution and aids in cell permeability
Applications:
  • Fluorescence microscopy
  • In vivo or ex vivo imaging
  • Cell-based assays
  • Flow cytometry/fluorescence-activated cell sorting (FACS)
Product Details:

DyLight 800-4xPEG Amine-Reactive Dye is activated with an N-hydroxysuccinimide (NHS) ester moiety to react with exposed N-terminal α-amino groups or the ε-amino groups of lysine residues to form stable amide bonds. Learn more about NHS ester chemistry.

Typical labeling reactions require the dye to first be dissolved in anhydrous dimethyl formamide (DMF) or another suitable organic solvent before adding a specific molar amount of dye to an amine-free buffer containing the protein to be labeled. However, the high solubility of DyLight Fluors permits protein solutions to be added directly to the labeling reagent.

Specifications

Quantity 1 mg
Product Type Reactive Dye
Solubility DMF (Dimethylformamide)
Content And Storage Store desiccated at -20°C.
Chemical Reactivity Amine
Reactive Moiety Active Ester, Succinimidyl Ester, NHS Ester
Label or Dye DyLight™ 800
Label Type DyLight Fluors
Product Line DyLight
Emission 794
Excitation 777
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.