missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Dye Primer/Matrix Standards [5FAM™, JOE™, TAMRA™, and ROX™ dyes]

Catalog No. 401114
Encompass
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
401114 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 401114 Supplier Applied Biosystems™ Supplier No. 401114

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Contains dye labeled standards used to generate multicomponent matrix. 1 tube of each dye

The standard can be used for Dye Primer cycle Sequencing and Dye Primer Sequenase Sequencing and on the 310, 3100/3100-Avant™ and 3130/XL Analyzers.

This Dye Primer Matrix Standards are used to generate the multicomponent matrix required when analyzing 5-FAM™, JOE™, TAMRA™ and ROX™-labeled DNA fragments.

Order Info

Each

Specifications

Content And Storage Contains dye labeled standards used to generate multicomponent matrix. 1 tube of each dye. Store the kit at 2 - 8°C. Do not freeze.
For Use With (Equipment) 310 Genetic Analyzer
For Use With (Application) Sequencing
Label or Dye FAM, JOE, ROX, TAMRA™
Product Type DNA Seqeuncing Size Standard
Quantity 1 Ea.
Shipping Condition Wet Ice

For Research Use Only. Not for use in diagnostic procedures.