missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DVL1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5149782
Description
DVL1 Polyclonal Antibody for Western Blot, IHC (P)
DVL1, the human homolog of the Drosophila dishevelled gene (dsh) encodes a cytoplasmic phosphoprotein that regulates cellproliferation, acting as a transducer molecule for developmentalprocesses, including segmentation and neuroblast specification.DVL1 is a candidate gene for neuroblastomatous transformation. TheSchwartz-Jampel syndrome and Charcot-Marie-Tooth disease type 2Ahave been mapped to the same region as DVL1. The phenotypes ofthese diseases may be consistent with defects which might beexpected from aberrant expression of a DVL gene during development.Specifications
| DVL1 | |
| Polyclonal | |
| Unconjugated | |
| DVL1 | |
| dishevelled 1; dishevelled 1 (homologous to Drosophila dsh); dishevelled segment polarity protein 1; dishevelled, dsh homolog 1; dishevelled, dsh homolog 1 (Drosophila); dishevelled-1; Dishevelled-1-like; DRS2; DSH homolog 1; DSH homolog 1-like; Dvl; Dvl1; dvl-1; DVL1L1; DVL1P1; MGC54245; mKIAA4029; RP5-890O3.5; segment polarity protein dishevelled homolog DVL-1 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 13542, 1855 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| O14640, P51141 | |
| DVL1 | |
| Recombinant Dishevelled, Dsh Homolog 1 (DVL1) (Arg150-Lys285). | |
| 200 μL | |
| Primary | |
| Human, Mouse | |
| Antibody |