missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DVL1 Polyclonal Antibody
GREENER_CHOICE

Goat Polyclonal Antibody

Supplier:  Invitrogen™ PA5142879

Catalog No. PIPA5142879


Only null left

Description

Description

This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:128,000.

DVL1, the human homolog of the Drosophila dishevelled gene (dsh) encodes a cytoplasmic phosphoprotein that regulates cellproliferation, acting as a transducer molecule for developmentalprocesses, including segmentation and neuroblast specification.DVL1 is a candidate gene for neuroblastomatous transformation. TheSchwartz-Jampel syndrome and Charcot-Marie-Tooth disease type 2Ahave been mapped to the same region as DVL1. The phenotypes ofthese diseases may be consistent with defects which might beexpected from aberrant expression of a DVL gene during development.
Specifications

Chemical Identifiers

DVL1
-20°C, Avoid Freeze/Thaw Cycles
Polyclonal
Liquid
IgG
Human
DVL1
dishevelled 1; dishevelled 1 (homologous to Drosophila dsh); dishevelled segment polarity protein 1; dishevelled, dsh homolog 1; dishevelled, dsh homolog 1 (Drosophila); dishevelled-1; Dishevelled-1-like; DRS2; DSH homolog 1; DSH homolog 1-like; Dvl; Dvl1; dvl-1; DVL1L1; DVL1P1; MGC54245; mKIAA4029; RP5-890O3.5; segment polarity protein dishevelled homolog DVL-1
DVL1
Primary
Ammonium sulfate precipitation
0.5 mg/mL
Immunohistochemistry (Paraffin)
Unconjugated
Goat
RUO
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
O14640
1855
Peptide with sequence KLPVAPERVTLAD-C (aa 20-32).
Antibody

Specifications

DVL1
Polyclonal
Unconjugated
DVL1
dishevelled 1; dishevelled 1 (homologous to Drosophila dsh); dishevelled segment polarity protein 1; dishevelled, dsh homolog 1; dishevelled, dsh homolog 1 (Drosophila); dishevelled-1; Dishevelled-1-like; DRS2; DSH homolog 1; DSH homolog 1-like; Dvl; Dvl1; dvl-1; DVL1L1; DVL1P1; MGC54245; mKIAA4029; RP5-890O3.5; segment polarity protein dishevelled homolog DVL-1
Goat
Ammonium sulfate precipitation
RUO
1855
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Immunohistochemistry (Paraffin)
0.5 mg/mL
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
O14640
DVL1
Peptide with sequence KLPVAPERVTLAD-C (aa 20-32).
100 μg
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions