missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Dutasteride, 99%

Catalog No. ac461512500
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
250 mg
Catalog No. Quantity
AC461512500 250 mg
AC461510010 1 g
2 options

Catalog No. AC461512500

Supplier: Thermo Scientific Chemicals 461512500

Only null left

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 164656-23-9
Molecular Formula C27H30F6N2O2
Molecular Weight (g/mol) 528.53
InChI Key JWJOTENAMICLJG-QWBYCMEYSA-N
Synonym dutasteride, avodart, avolve, duagen, unii-o0j6xjn02i, avodart tn, o0j6xjn02i, dutasteride jan/usan/inn, 5alpha,17beta-n-2,5-bis trifluoromethyl phenyl-3-oxo-4-azaandrost-1-ene-17-carboxamide, dutasteride usan
PubChem CID 6918296
ChEBI CHEBI:521033
IUPAC Name (1S,3aS,3bS,5aR,9aR,9bS,11aS)-N-[2,5-bis(trifluoromethyl)phenyl]-9a,11a-dimethyl-7-oxo-1,2,3,3a,3b,4,5,5a,6,9b,10,11-dodecahydroindeno[5,4-f]quinoline-1-carboxamide
SMILES CC12CCC3C(C1CCC2C(=O)NC4=C(C=CC(=C4)C(F)(F)F)C(F)(F)F)CCC5C3(C=CC(=O)N5)C

Specifications

Melting Point 245.0°C
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Quantity 250 mg
Solubility Information Solubility in water: insoluble.
Formula Weight 528.53
Percent Purity 99%
Chemical Name or Material Dutasteride
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May damage the unborn child.
Suspected of damaging fertility.
Harmful to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement Obtain special instructions before use. Wear protective gloves/protective clothing/eye protection/face protection. IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only