CAS RN 164656-23-9
IUPAC Name:
(4aR,6aS,7S,9aS,9bS,11aR)-N-[2,5-bis(trifluoromethyl)phenyl]-4a,6a-dimethyl-2-oxo-1H,2H,4aH,4bH,5H,6H,6aH,7H,8H,9H,9aH,9bH,10H,11H,11aH-indeno[5,4-f]quinoline-7-carboxamide
Synonyms:
dutasteride
Molecular Weight (g/mol):
528.54
Molecular Formula:
C27H30F6N2O2
InChi Key:
JWJOTENAMICLJG-VYZSUTEISA-N
SMILES:
C[C@]12CCC3[C@@H](CC[C@H]4NC(=O)C=C[C@]34C)[C@@H]1CC[C@@H]2C(=O)NC1=CC(=CC=C1C(F)(F)F)C(F)(F)F
1
–
1
of
1
results
1
Thermo Scientific Chemicals Dutasteride, 99%
CAS: 164656-23-9 Molecular Formula: C27H30F6N2O2 Molecular Weight (g/mol): 528.53 InChI Key: JWJOTENAMICLJG-QWBYCMEYSA-N Synonym: dutasteride,avodart,avolve,duagen,unii-o0j6xjn02i,avodart tn,o0j6xjn02i,dutasteride jan/usan/inn,5alpha,17beta-n-2,5-bis trifluoromethyl phenyl-3-oxo-4-azaandrost-1-ene-17-carboxamide,dutasteride usan PubChem CID: 6918296 ChEBI: CHEBI:521033 IUPAC Name: (1S,3aS,3bS,5aR,9aR,9bS,11aS)-N-[2,5-bis(trifluoromethyl)phenyl]-9a,11a-dimethyl-7-oxo-1,2,3,3a,3b,4,5,5a,6,9b,10,11-dodecahydroindeno[5,4-f]quinoline-1-carboxamide SMILES: CC12CCC3C(C1CCC2C(=O)NC4=C(C=CC(=C4)C(F)(F)F)C(F)(F)F)CCC5C3(C=CC(=O)N5)C