missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bio-Serv™ Dustless Precision Pellets™ Purified Rodent Diet
Precise size and weight pellets generally used in nutritional and behavioral studies
$288.54 - $601.88
Specifications
| Product Type | Rodent Diet |
|---|---|
| Calories | 3.6 |
| For Use With (Application) | Nutritional and behavioral studies |
| Particle Shape | Pellet |
| Species | Rodent |
| Catalog Number | Mfr. No. | Diameter (English) | Diameter (Metric) | Quantity | Flavor | Price | Quantity | ||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Diameter (English) | Diameter (Metric) | Quantity | Flavor | Price | Quantity | ||
14726596
|
Bio-Serv™
F0299 |
0.14 in. | 0.036 cm | 0.045 g | Chocolate |
Each for $329.06
|
|
||
14726595
|
Bio-Serv™
F05301 |
0.1 in. | 0.025 cm | 0.02 g | Chocolate |
Each for $356.33
|
|
||
14726750
|
Bio-Serv™
F0021 |
0.14 in. | 0.036 cm | 0.045 g | No Added Flavor |
Each for $294.21
|
|
||
14726589
|
Bio-Serv™
F0071 |
0.1 in. | 0.025 cm | 0.02 g | No Added Flavor |
Each for $288.54
|
|
||
14726592
|
Bio-Serv™
F0075 |
0.3 in. | 0.076 cm | 0.3 g | No Added Flavor |
Each for $601.88
|
|
||
14726594
|
Bio-Serv™
F0078 |
0.45 in. | 0.114 cm | 1 g | No Added Flavor |
Each for $314.69
|
|
||
Description
Precise size and weight pellets generally used in nutritional and behavioral studies
- Nutritionally complete purified formula
- Dustless and fracture free—superior performance in automatic feeders
- Every batch tested for hardness, weight and height
- Nutritionally assayed
Ingredients: Sucrose. Dextrose, Casein, Corn Oil, Mineral Mix, Cellulose, Corn Syrup, Calcium Silicate, Vitamin Mix, Magnesium Stearate, Choline Bitartrate, DL-Methionine, L-Cystine, Ascorbic Acid, Vitamin E Acetate, tBHQ, Chocolate Flavoring (F05301 and F0299 only)
Specifications
| Rodent Diet | |
| Nutritional and behavioral studies | |
| Rodent |
| 3.6 | |
| Pellet | |
| Cool dry place |