missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bio-Serv™ Dustless Precision Pellets™ Purified Rodent Diet
Precise size and weight pellets generally used in nutritional and behavioral studies
Supplier: Bio-Serv™ F0078
Description
Precise size and weight pellets generally used in nutritional and behavioral studies
- Nutritionally complete purified formula
- Dustless and fracture free—superior performance in automatic feeders
- Every batch tested for hardness, weight and height
- Nutritionally assayed
Ingredients: Sucrose. Dextrose, Casein, Corn Oil, Mineral Mix, Cellulose, Corn Syrup, Calcium Silicate, Vitamin Mix, Magnesium Stearate, Choline Bitartrate, DL-Methionine, L-Cystine, Ascorbic Acid, Vitamin E Acetate, tBHQ, Chocolate Flavoring (F05301 and F0299 only)
Specifications
| Rodent Diet | |
| Sucrose. Dextrose, Casein, Corn Oil, Mineral Mix, Cellulose, Corn Syrup, Calcium Silicate, Vitamin Mix, Magnesium Stearate, Choline Bitartrate, DL-Methionine, L-Cystine, Ascorbic Acid, Vitamin E Acetate, tBHQ | |
| Nutritional and behavioral studies | |
| 1 g | |
| Cool dry place | |
| 0.114 cm |
| 3.6 | |
| No Added Flavor | |
| Pellet | |
| Rodent | |
| 0.45 in. |