missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ DuraPorter™ XL Tube Insert

DuraPorter™ XL Tube Rack Insert designed to fit DuraPorter™ XL High-Capacity Sample Transport.

Supplier:  Fisherbrand™ 120961

 View more versions of this product

Catalog No. 17183977



Only null left

Description

Description

  • Designed to fit DuraPorter™ XL High-Capacity Sample Transport
  • Thermoformed PETG
  • Tube Rack Inserts can be used separately
  • Lid removes completely for easy cleaning
  • Insert deep enough to be filled with ice
  • Holds 100 tubes with diameter up to 18 mm, most 10 mL blood tubes and 15 mL conical tubes
Specifications

Chemical Identifiers

Not Autoclavable
Thermoformed PETG
Rectangular, Rounded Corners
Non-sterile
Tube Holder
Clear
1
Yes, When Empty
13.1 x 14.3 x 5.55 in.
None

Specifications

Not Autoclavable
Blood Tubes (Small)
1
Yes, When Empty
13.1 x 14.3 x 5.55 in.
None
Clear
Thermoformed PETG
Rectangular, Rounded Corners
Non-sterile
Tube Holder
12 x 14
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions