Learn More
DWK Life Sciences DURAN™ YOUTILITY Laboratory Bottles GL45
Help to make laboratory work easier, safer, more economical and fun
Supplier:  DWK Life Sciences 218813653
 View more versions of this product
Catalog No. 09841066
Only null left
Description
Description
  • Specially shaped gripping zones on both sides of the bottle for easier and safer handling
  • Opening or closing of the bottle is faster, and fully compatible with DIN GL 45 closures and other accessories
  • Slimmer YOUTILITY bottle shape allows a more optimal use of limited space in autoclaves and laboratory refrigerators
  • Pre-defined labelling area is compatible with DURAN™ self-adhesive YOUTILITY bottle labels
  • Nominal volume is shown at the top of the easy-to-read graduation scale for fast determination of the volumes
  • Sequentially sized bottles are available in four volumes
  • Moulded from the DURAN™ borosilicate 3.3 pharmacopeia type 1 neutral glass
  • Retrace Code for full product traceability
  • Each bottle is supplied as a complete system, with a pouring ring (cyan polypropylene) and a GL 45 cap (cyan polypropylene)
Specifications
Chemical Identifiers
Clear
Glass
Specifications
250 mL
Glass
66 mm
Clear
158 mm
Product Suggestions
Product Suggestions