missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences DURAN™ Screw Caps for GL S 80™ Wide Neck Glass Bottles
High leak tightness through use of PTFE coated silicone seal. More chemically resistant than PP screw cap.
$47.65
Specifications
| Color | Red |
|---|---|
| Material | PBT |
| For Use With (Equipment) | GLS 80 wide neck glass bottles |
| Catalog Number | Mfr. No. | Thread Size (Metric) | Product Type | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Thread Size (Metric) | Product Type | Price | Quantity & Availability | ||||
|
09841257
|
DWK Life Sciences
1170682 |
GL 32 | Screw Cap with PTFE/Silicone-protected Seal Liner |
Pack of 2 Each for $47.65
|
|
||||
Description
- Matching ETFE pouring ring is also available, permitting clean, drip-free use
Specifications
| Red | |
| PBT | |
| GLS 80 wide neck glass bottles |