missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences DURAN™ Screw Caps for GL S 80™ Wide Neck Glass Bottles

Catalog No. 09841257
Change view
Click to view available options
Thread Size (Metric):
GL 32
Product Type:
Screw Cap with PTFE/Silicone-protected Seal Liner
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Thread Size (Metric) Product Type
09841257 GL 32 Screw Cap with PTFE/Silicone-protected Seal Liner
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09841257 Supplier DWK Life Sciences Supplier No. 1170682

High leak tightness through use of PTFE coated silicone seal. More chemically resistant than PP screw cap.

  • Matching ETFE pouring ring is also available, permitting clean, drip-free use

Specifications

Color Red
Material PBT
Thread Size (Metric) GL 32
Product Type Screw Cap with PTFE/Silicone-protected Seal Liner
For Use With (Equipment) GLS 80 wide neck glass bottles