missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences™ DURAN™ protect+ Graduated Laboratory Bottles with DIN 168-1 Thread
GREENER_CHOICE

Multi-functional clear glass laboratory bottles with protective safety coating.

Supplier:  DWK Life Sciences™ 218054487

 View more versions of this product

Catalog No. 13299181

Only null left

Description

Description

DURAN™ GL ISO bottles have become ideal multi-functional laboratory bottles since their introduction in 1972. The protect+ safety-coated version of the bottles combines the best of both worlds: virtually inert USP/EP Type I borosilicate glass 3.3 on the inside and a protective safety plastic layer on the outside. The bottles are molded from high-purity, clear glass or externally ambered clear glass that is then coated with a translucent polyamide PA 12 coating to reduce the hazards of broken glass and liquid splashes in the event of breakage.

A combination of high-performance materials and robust design provides a cost-effective usable life. The bottles feature an easy-to-read graduated volume scale. There is a Retrace Code printed on each bottle for manufacturing lot traceability. The ambered DURAN™ glass bottle versions ensure that light sensitive contents are protected from the effects of photodegradation. The protection against ultra-violet light meets international USP/EP standards. All the protect+ coated bottles are supplied with push-on protective covers to protect the inside of the bottles from contamination during distribution. Matching screw closures and pouring rings can be ordered separately. Available in a wide range of nominal bottle capacities from 25 mL to 5 L, and neck sizes GL 25 to GLS 80™ wide mouth.

  • Manufactured from polyamide-coated DURAN™ borosilicate glass 3.3
  • DURAN™ is a neutral glass, classified as a Glass Type I according to USP, EP (3.2.1) and JP, and meets ASTM E438, Type I, Class A requirements
  • Reduce the hazards of broken glass and liquid splashes in the event of breakage
  • External polymer coating provides scratch, leak* and splinter* protection (*only applies to bottles 2000 mL or less)
  • Polyamide 12 safety coating has a high chemical resistance
  • Protective coating provides an enhanced safer grip
  • Coating is Cleanroom certified Fraunhofer IPA TESTED DEVICE™
  • Certified FDA and EU food contact safe coating material
  • Safe storage and shipping of bulk, high-potency active pharmaceutical ingredients (HPAPIs) powders
  • Supplied with push-on dust cover
  • Coating process does not use any volatile organic compounds (VOCs) or isocyanates
Specifications

Chemical Identifiers

Clear
Borosilicate Glass (3.3)
No
Wide
Laboratory Bottle
Preparation of media and reagents, safe storage and shipping of bulk, high-potency active pharmaceutical ingredients (HPAPIs), sample or harvest collection container, culturing vessel, storage of media and reagents, transport and shipping, Liquid handling reservoir
Non-sterile
Yes
Round
Not Autoclavable

Specifications

500 mL
Clear
Borosilicate Glass (3.3)
Non-sterile
GL 45
No
Wide
181 mm
45
Not Autoclavable
16.9 oz.
Preparation of media and reagents, safe storage and shipping of bulk, high-potency active pharmaceutical ingredients (HPAPIs), sample or harvest collection container, culturing vessel, storage of media and reagents, transport and shipping, Liquid handling reservoir
10 Bottles
0.356 kg
CoC
Yes
Round
Laboratory Bottle
86 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions