Learn More
DWK Life Sciences™ DURAN™ protect+ Graduated Amber Laboratory Bottle with DIN 168-1 Thread

Multi-functional ambered glass laboratory bottles with UV protection and a protective safety coating.
$1,752.62 - $3,722.62
Specifications
| Color | Amber |
|---|---|
| For Use With (Application) | Preparation of light-sensitive media and reagents, safe storage and shipping of bulk, light-sensitive, high-potency active pharmaceutical ingredients (HPAPIs) powders, sample or harvest collection container, sulturing vessel, storage of light-sensitive media and reagents, transport and shipping |
| Material | Borosilicate Glass (3.3) |
| Sterility | Non-sterile |
| Certifications/Compliance | CoC |
| Catalog Number | Mfr. No. | Capacity (Metric) | Thread Size (Metric) | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity (Metric) | Thread Size (Metric) | Price | Quantity & Availability | ||||
13299176
![]() |
DWK Life Sciences™
218061482 |
25 mL | GL 25 |
Case of 10 Each for $1,752.62
|
|
||||
13299175
![]() |
DWK Life Sciences™
218061782 |
50 mL | GL 32 |
Case of 10 Each for $1,772.41
|
|
||||
13299174
![]() |
DWK Life Sciences™
218062487 |
100 mL | GL 45 |
Case of 10 Each for $1,772.41
|
|
||||
13299173
![]() |
DWK Life Sciences™
218063689 |
250 mL | GL 45 |
Case of 10 Each for $2,309.56
|
|
||||
13299172
![]() |
DWK Life Sciences™
218064488 |
500 mL | GL 45 |
Case of 10 Each for $2,865.71
|
|
||||
13299171
![]() |
DWK Life Sciences™
218065484 |
1 L | GL 45 |
Case of 10 Each for $3,722.62
|
|
||||
13299170
![]() |
DWK Life Sciences™
218066386 |
2 L | GL 45 |
Case of 6 Each for $3,439.56
|
|
||||
13299169
![]() |
DWK Life Sciences™
218067382 |
5 L | GL 45 |
Case for $2,096.41
|
|
||||
Description
DURAN™ GL ISO bottles have become ideal multi-functional laboratory bottles since their introduction in 1972. The protect+ safety-coated version of the bottles combines the best of both worlds: virtually inert USP/EP Type I borosilicate glass 3.3 on the inside and a protective safety plastic layer on the outside. The bottles are molded from high-purity, clear glass or externally ambered clear glass that is then coated with a translucent polyamide PA 12 coating to reduce the hazards of broken glass and liquid splashes in the event of breakage.
A combination of high-performance materials and robust design provides a cost-effective usable life. The bottles feature an easy-to-read graduated volume scale. There is a Retrace Code printed on each bottle for manufacturing lot traceability. The ambered DURAN™ glass bottle versions ensure that light sensitive contents are protected from the effects of photodegradation. The protection against ultra-violet light meets international USP/EP standards. All the protect+ coated bottles are supplied with push-on protective covers to protect the inside of the bottles from contamination during distribution. Matching screw closures and pouring rings can be ordered separately. Available in a wide range of nominal bottle capacities from 25 mL to 5 L, and neck sizes GL 25 to GLS 80™ wide mouth.
- Manufactured from ambered borosilicate glass 3.3 and polyamide
- Standardized GL-thread compatible with matching cap system for particularly tight sealing and simple, clean pouring out
- The protect+ safety coating provides scratch, leak* and splinter* protection (*only applies to bottles 2000 mL or less)
- UV protection USP (Spectral Transmission) and EP (3.2.1) compliant
- Less than 10% spectral transmission between 290 nm and 450 nm
- The only external application of the amber color and safety coating ensures that internal surfaces have standard DURAN™ glass properties
- Supplied as safety coated glass bottle only (screw caps and pouring rings must be ordered separately)
- Easy-to-read scale with graduation marks to estimate the content
- The 25 mL has a specially shaped glass edge for improved pouring out whereby an additional plastic pouring ring is not required
- Safe storage and shipping of bulk, high-potency active pharmaceutical ingredients (HPAPIs) powders
- Coating process does not use any volatile organic compounds (VOCs) or isocyanates
Specifications
| Amber | |
| Borosilicate Glass (3.3) | |
| CoC | |
| Yes | |
| Round | |
| Not Autoclavable |
| Preparation of light-sensitive media and reagents, safe storage and shipping of bulk, light-sensitive, high-potency active pharmaceutical ingredients (HPAPIs) powders, sample or harvest collection container, sulturing vessel, storage of light-sensitive media and reagents, transport and shipping | |
| Non-sterile | |
| No | |
| Wide | |
| Laboratory Bottle |