Learn More
DWK Life Sciences DURAN™ Original GL 45 Laboratory Bottle, with High Temperature Screw cap, Clear, with DIN 168-1 Thread, Clear, Graduated
Description
DURAN™ Original GL laboratory bottles have become the gold standard of multifunctional laboratory bottles since their introduction in 1972. The bottles are moulded from high-purity, clear, USP/EP Type I DURAN™ borosilicate glass 3.3, renowned for its excellent thermal performance, and high resistance to chemical attack. A combination of high-performance materials and robust ISO 4796-1 design provides a long working life. The bottles feature an easy-to-read graduated volume scale, and marking field in permanent white enamel. Permanent Retrace Code on each bottle for manufacturing lot traceability. DURAN Original GL 45 laboratory bottles are available complete with high temperature screw caps and pouring rings. The red polybutylene terephthalate (PBT) GL 45 threaded caps with PTFE faced silicone cap liner and ETFE pouring rings offer greater thermal and chemical resistance than the equivalent polypropylene components. All components are fully autoclavable at 121 or 134°C and suitable for dry heat sterilization up to 180°C. All components are fully compliant with current international food contact regulations such as (EU) No 10/2011.
- Supplied complete with red polybutylene terephthalate (PBT) GL 45 threaded cap with PTFE faced silicone liner and ETFE pouring ring for drip free pouring.
- Supplied complete with high temperature PBT screw caps and ETFE pouring rings.
- The bottles feature an easy-to-read graduated volume scale, and marking field in permanent white enamel.
- Permanent Retrace Code on each bottle for full manufacturing lot traceability.
- Suitable for long-term storage.
- All components are fully autoclavable at 121 or 134°C.
- All components are suitable for dry heat sterilization up to 180°C.
Specifications
Specifications
| Capacity (Metric) | 500 mL |
| For Use With (Application) | Suitable for dry heat sterilization; Autoclaving of liquid media; Storage of corrosive reagents; Sampling; Storage of media and reagents; Transport and shipping |
| Includes | With high temperature resistant PBT screw cap and ETFE pouring ring |
| Material | Borosilicate Glass (3.3) |
| Quantity | 10/Pk. |
| Thread Size (Metric) | GL 45 |
| Closure Material | PBT with ETFE Pouring Ring |
| Diameter (Metric) | 86 mm |
| Autoclavable | Autoclavable |