Learn More
DWK Life Sciences DURAN™ GLS 80™ Laboratory Bottle, Wide Mouth, Clear, GLS 80™ Thread, Graduated, Bottle Only
Wide mouth, multifunctional glass laboratory bottles.
$418.53 - $3,439.65
Specifications
| For Use With (Application) | Autoclaving of media and reagents; Preparation of media and reagents; Sample collection; Culturing vessel; Storage of media and reagents; Transport and shipping; Liquid handling reservoir |
|---|---|
| Includes | Without screw cap and pouring ring from PP (blue) |
| Material | Borosilicate Glass (3.3) |
| Thread Size (Metric) | GLS 80 |
| Closure Material | Polypropylene |
| Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity & Availability | |||||
|
09841208
|
DWK Life Sciences
1178425 |
2000 mL |
Case of 10 Each for $1,826.82
|
|
|||||
|
09841206
|
DWK Life Sciences
1178392 |
500 mL |
Case of 10 Each for $641.56
|
|
|||||
|
02900154
|
DWK Life Sciences
218606904 |
3500 mL |
Case for $778.94
|
|
|||||
|
09841209
|
DWK Life Sciences
1178426 |
5000 mL |
Each for $912.24
|
|
|||||
|
09841210
|
DWK Life Sciences
1178427 |
10,000 mL |
Case for $1,565.24
|
|
|||||
|
09841211
|
DWK Life Sciences
1178428 |
20,000 mL |
Case for $3,439.65
|
|
|||||
|
09841212
|
DWK Life Sciences
218603607 |
250 mL |
Case of 10 Each for $418.53
|
|
|||||
Description
DURAN™ wide mouth GLS 80™ laboratory bottles are moulded from high-purity, clear, USP/EP Type I borosilicate glass 3.3, renowned for its excellent thermal performance, and high resistance to chemical attack. A combination of high-performance materials and robust ISO 4796-1 design provides a long working life. The bottles feature an easy-to-read graduated volume scale, and marking field in permanent white enamel. Permanent Retrace Code on each bottle for manufacturing lot traceability. DURAN™ wide mouth GLS 80™ are supplied complete with blue polypropylene cap and drip-free pouring ring for use up to 140°C. All components are fully autoclavable at 121 or 134°C.
- Easy access to the bottle contents due to the wide diameter of the bottle neck
- Special thread design for rapid opening and closing in less than a turn
- Optimum filling and pouring and particularly easy cleaning
- Spoons, spatulas and tweezers can be manipulated with ease
- Even large-volume funnels can be inserted into the bottles without any difficulty
- Stability due to the large base area
- Easy-to-read scale with graduation marks to estimate the content
- Fired-on, highly durable white ceramic
- Complete with blue screw cap (PP, integral lip seal) and pouring ring (PP) for drip-free pouring and clean, safe working
Specifications
| Autoclaving of media and reagents; Preparation of media and reagents; Sample collection; Culturing vessel; Storage of media and reagents; Transport and shipping; Liquid handling reservoir | |
| Borosilicate Glass (3.3) | |
| Polypropylene |
| Without screw cap and pouring ring from PP (blue) | |
| GLS 80 | |
| Autoclavable |