missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences DURAN™ GL 45 Bottle

Catalog No. 09841316
Change view
Click to view available options
Capacity (Metric):
2000 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
09841316 2000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09841316 Supplier DWK Life Sciences Supplier No. 219900701

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

DURAN™ GL 45 Bottle. The DURAN™ GL 45 Bottle is ideal for storage and delivery of HPLC mobile phases and solvents. Borosilicate 3.3 glass offers low extractables, GL 45 threads fit standard HPLC caps, and a side neck allows solvent filling without disturbing the main delivery cap.

  • Ideal for the storage and delivery of high-performance liquid chromatography (HPLC) mobile phases and solvents.
  • Increasing the number of GL 45 inlets on the bottles adds convenience: One additional side neck allows additional solvent to be added without disturbing the main delivery cap.
  • Convenient – Side neck allow solvent filling without main delivery cap removal
  • Safer – Capped bottles reduce the risk of volatile organic compounds (VOC) escape and spillage
  • Compatible - GL 45 threads fit standard HPLC connection caps
  • Manufactured from the material of choice for HPLC solvent bottles: borosilicate 3.3 glass offering a very low inorganic ion extractables profile
  • Clear DURAN™ is a borosilicate glass type 3.3 as specified by the international standard ISO 3585, and is classified as a Type I, Class A low-expansion borosilicate glass as defined by ASTM-E438-1992
  • Useful 2 Liter Capacity
  • Retrace Code, and certificate for manufacturing lot traceability
  • Durable white enamel volume graduations

Specifications

Capacity (Metric) 2000 mL
Cleaning Procedure Lab Washer Compatible
Color Clear
Diameter (Metric) Inner Neck 30 mm
For Use With (Application) Mobile Phase Reservoirs for HPLC, Reagent Supply to Liquid Handling Systems and Analyzers
Includes GL 45 Side Neck, Two GL 45 inlets
Material Borosilicate Glass (3.3)
Max. Temperature (Metric) 500°C
Quantity 1
Screw Color Blue
Screw Material Polypropylene
Sterility Non-sterile
Weight (Metric) 950 g
Width (Metric) 136 mm
Thread Size (Metric) GL 45
Closure Color Blue
Graduations 400 to 1800
Nominal Capacity 2000 mL
Closure Included Yes
Graduated Yes
Mouth Narrow
Height (Metric) 265.5 mm
Closure Material Polypropylene
Diameter (Metric) 136 mm
Closure Type Screw Cap
Autoclavable Autoclavable
Capacity (English) 68 oz.
Shape Cylindrical
Certifications/Compliance DIN ISO 3585, Glass Type I according to the requirements of the current United States pharmacopoeia USP <660> Containers - Glass, European pharmacopoeia EP (3.2.1) Glass Containers for Pharmaceutical Use, Japanese pharmacopoeia JP <7.01> Test for Glass Containers for injections, This batch certificate confirms that ASTM Class A error limits required by USP are met. The certificate records the mean value, the standard deviation of the batch and the date of issue.
Disposable Reusable
Product Type Reservoir Bottle
Show More Show Less