Learn More
DWK Life Sciences Flexible modular system with four different tubing diameters (1.6 mm; 3.0 mm; 3.2 mm and 6.0 mm) can be connected. Sterile pressure equalization is possible through use of syringe filter. Unused ports can be provided with a blind cap.
Flexible modular system with four different tubing diameters (1.6 mm; 3.0 mm; 3.2 mm and 6.0 mm) can be connected. Sterile pressure equalization is possible through use of syringe filter. Unused ports can be provided with a blind cap.
Supplier:  DWK Life Sciences 1156292
 View more versions of this product
Catalog No. 09-841-113
Only null left
Explore available promotions
Description
Description
Flexible modular system with four different tubing diameters (1.6 mm; 3.0 mm; 3.2 mm and 6.0 mm) can be connected. Sterile pressure equalization is possible through use of syringe filter. Unused ports can be provided with a blind cap.
  • Materials: PP and PTFE
  • Typical applications: safe transfer of liquid media within a closed and sterile system (evaporation is reduced)
Specifications
Specifications
Screw Cap
19.7 mm
Polybutylene
For safe transfer of liquid media within a closed and sterile system
GL14
Autoclavable
Red
Duran™ Laboratory Glass Bottles
Product Suggestions
Product Suggestions