missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences DURAN™ Aspirator Laboratory Bottle, GL 45 Thread Neck, GL 32 Thread Side Arm
Aspirator laboratory bottle with screw closure and PTFE key stopcock
$1,706.68
Specifications
| For Use With (Application) | Storage and dispensing of liquids and reagents |
|---|---|
| Material | Borosilicate Glass (3.3), Silicone, PTFE |
| No. per Pack | 1/Pk. |
| Neck Finish | Thread Size GL 45 |
| Autoclavable | Autoclavable |
Description
Aspirator laboratory bottle with screw closure, complete with PTFE key stopcock and GL 45 red PBT cap.
- Supplied ready to use
- DURAN™ glass and PTFE key stopcock outlet
- Made of virtually inert, chemically resistant DURAN™ glass
- Fully compatible with all the GL 45 threads and therefore highly versatile
- Tabulator with GL 32 thread
- Easy-to-read scale with graduation marks to estimate the content
- Fired-on, highly durable white ceramic
Specifications
| Storage and dispensing of liquids and reagents | |
| 1/Pk. | |
| Autoclavable |
| Borosilicate Glass (3.3), Silicone, PTFE | |
| Thread Size GL 45 |