Learn More
DuPont™ Tyvek™ 400 Lab Coat
Limited-use lab coats offer easy donning and doffing and provide an ideal balance of protection, durability, and comfort.
Supplier:  DuPont™ TY212SWHMD0030VP
 View more versions of this product
Catalog No. 19046143B
Only null left
Explore available promotions
Alert:
Garments made of Tyvek should not be used around heat, flames, sparks or in potentially flammable or explosive environments.
Description
Description
Limited-use lab coats offer easy donning and doffing and provide an ideal balance of protection, durability, and comfort.
  • Labcoat offers front snap closures for easy donning and doffing
  • Provides enhanced frontal protection
  • Laydown collar
  • Open wrists
  • Two front pockets for holding writing instruments, small tools or parts
  • Manufactured under specifications that do not contain natural rubber latex
Compliance
Meets ANSI/ISEA 101-1996 standards.
Specifications
Specifications
General Purpose Lab Coat
Unisex
White
Open
Non-sterile
2
TY212SWH
35.25 to 38.75 in.
Individually Packaged for PPE Vending Machines
32 in.
81.28 cm
Medium
Tyvek
Snaps
Limited-use
Below Hip-length
Lay Down
Serged
Lead and Asbestos Abatement, Remediation, General Maintenance, Spray Painting, Cleaning
34 in.
86.36 cm
Product Suggestions
Product Suggestions