missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ Tyvek™ IsoClean™ Series 447 Boot Covers
Gripper™ soles resists slipping on slick surfaces
Supplier: DuPont™ IC447SWHXL0100CS
Description
Tyvek IsoClean fabric is antistatic and low linting and provides an excellent barrir to dry particulates, nonhazardous liquids and microorganisms.
- Coarsely textured interior to help prevent movement inside
- PVC Gripper soles provide excellent barrier to liquid penetration
- 18 in. (45.7cm) high Tyvek uppers with elastic leg and ankle openings
- Serged seams
- White
- Available clean processed and gamma sterilized or bulk packaged
Order Info
Clean and sterile covers are individually packaged in opaque bag and placed in double transparent poly liners in and ISO Class 4 cleanroom. Bulk covers are bulk packaged in double transparent poly liners.
Specifications
| X-Large | |
| Yes | |
| Sterile | |
| Serged | |
| 50 Pairs | |
| 38.7 cm | |
| 45.7 cm |
| Tyvek | |
| White | |
| Elastic Top and Ankles | |
| Ideal To Provide An Excellent Barrier To Dry Particles, Non-hazardous Liquids And Microorganisms | |
| 15.25 in. | |
| 18 in. |