missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Tyvek™ IsoClean™ Series 447 Boot Covers

Catalog No. p-4591601
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
Length (Metric):
27.3 cm
33.7 cm
38.7 cm
43.8 cm
Sterility:
Non-sterile
Sterile
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Length (Metric) Sterility
19039728D X-Large 38.7 cm Non-sterile
19039852 Large 33.7 cm Sterile
19039851 Medium 27.3 cm Sterile
19039853 X-Large 38.7 cm Sterile
19039854 2X-Large 43.8 cm Sterile
19039870 Medium 27.3 cm Non-sterile
19039871 Large 33.7 cm Non-sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 19039728D Supplier DuPont™ Supplier No. IC447SWHXL01000B
Only null left

Gripper™ soles resists slipping on slick surfaces

Tyvek IsoClean fabric is antistatic and low linting and provides an excellent barrir to dry particulates, nonhazardous liquids and microorganisms.

  • Coarsely textured interior to help prevent movement inside
  • PVC Gripper soles provide excellent barrier to liquid penetration
  • 18 in. (45.7cm) high Tyvek uppers with elastic leg and ankle openings
  • Serged seams
  • White
  • Available clean processed and gamma sterilized or bulk packaged

Order Info

Clean and sterile covers are individually packaged in opaque bag and placed in double transparent poly liners in and ISO Class 4 cleanroom. Bulk covers are bulk packaged in double transparent poly liners.

Specifications

Size X-Large
Material Tyvek
Latex Free Yes
Color White
Sterility Non-sterile
Closure Type Elastic Top and Ankles
Seam Type Serged
For Use With (Application) Ideal To Provide An Excellent Barrier To Dry Particles, Non-hazardous Liquids And Microorganisms
Packaging Quantity 50 Pairs
Length (English) 15.25 in.
Length (Metric) 38.7 cm
Height (English) 18 in.
Height (Metric) 45.7 cm
Show More Show Less