missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ Tyvek™ IsoClean™ Zipper Front Cleanroom Frocks
Frock provides an inherent barrier to particles, microorganisms and non-hazardous light liquid splash. Bulk packaged in double transparent poly liners.
Supplier: DuPont™ IC264SWHLG00300B
Alert:
Should not be worn around open flame, heat source, sparks or ignition sources.
Description
Frock provides an inherent barrier to particles, microorganisms and non-hazardous light liquid splash. ulk packaged in double transparent poly liners.
- Serged seams have multiple interlocking threads that are sewn around the raw edges of garment material to create a strong, stress-resistant seam
- Elastic openings for tighter fit at wrist
- Bound neck for lower particle shedding
- Front zipper closure for easy donning and doffing
- Raglan sleeve design for greater range of motion and comfort
- A-line silhouette
Specifications
| Controlled Environments Frock | |
| Tyvek | |
| Non-sterile | |
| Elastic | |
| Bound | |
| 91.44 to 96.52 cm | |
| Bulk Packaged | |
| 116.21 cm | |
| 97.15 cm |
| Large | |
| White | |
| Zipper | |
| Serged | |
| 36 to 38 in. | |
| Raglan Sleeve | |
| 45.75 in. | |
| 38.25 in. | |
| Class 1000 (ISO 6) |