missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ Tyvek™ IsoClean™ Zipper Front Cleanroom Frocks

Description
Frock provides an inherent barrier to particles, microorganisms and non-hazardous light liquid splash. ulk packaged in double transparent poly liners.
- Serged seams have multiple interlocking threads that are sewn around the raw edges of garment material to create a strong, stress-resistant seam
- Elastic openings for tighter fit at wrist
- Bound neck for lower particle shedding
- Front zipper closure for easy donning and doffing
- Raglan sleeve design for greater range of motion and comfort
- A-line silhouette
Specifications
Specifications
| Product Type | Controlled Environments Frock |
| Size | Medium |
| Material | Tyvek |
| Color | White |
| Sterility | Non-sterile |
| Closure Type | Zipper |
| Cuff Style | Elastic |
| Seam Type | Serged |
| Collar Type | Bound |
| Size (English) Chest | 32 to 36 in. |
| Show More |