missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Tyvek™ IsoClean™ Zipper Front Cleanroom Frocks

Frock provides an inherent barrier to particles, microorganisms and non-hazardous light liquid splash. Bulk packaged in double transparent poly liners.

Supplier:  DuPont™ IC264SWH3X00300B

 View more versions of this product

Catalog No. 17-987-100


Only null left
Explore available promotions

Alert:

Should not be worn around open flame, heat source, sparks or ignition sources.


Description

Description

Frock provides an inherent barrier to particles, microorganisms and non-hazardous light liquid splash. ulk packaged in double transparent poly liners.

  • Serged seams have multiple interlocking threads that are sewn around the raw edges of garment material to create a strong, stress-resistant seam
  • Elastic openings for tighter fit at wrist
  • Bound neck for lower particle shedding
  • Front zipper closure for easy donning and doffing
  • Raglan sleeve design for greater range of motion and comfort
  • A-line silhouette
Specifications

Specifications

Controlled Environments Frock
Tyvek
Non-sterile
Elastic
Bound
109.22 to 116.84 cm
Bulk Packaged
139.7 cm
112.39 cm
3X-Large
White
Zipper
Serged
43 to 46 in.
Raglan Sleeve
55 in.
44.25 in.
Class 1000 (ISO 6)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions