Learn More
DuPont™ Tyvek™ IsoClean™ Series 181 Coveralls
Protects against micro-organisms and particulates in ISO 4 Class 10 cleanroom environments.
Supplier:  DuPont™ IC181SWH7X002500
 View more versions of this product
Catalog No. 17-978-152
Only null left
Explore available promotions
Description
Description
Clean-processed garments offer lowest level of particle shedding within DuPont product portfolio
  • Clean processed
  • Serged seams
  • Elastic wrists and ankles
  • Zipper closure
  • Use in controlled environments with high standards for particle and microbiological contamination control
  • Pharmaceutical, biotech, medical device and electronic settings
Specifications
Specifications
Cleanroom Coveralls
Unisex
White
Elastic
Zipper
Non-sterile
Clean Processed
65 to 68 in.
165.1 to 172.7 cm
41.75 in.
Bulk
7X-Large
Tyvek Flashspun Polyethylene
Single-use
Elastic
Serged
Laydown
Yes
35.5 in.
90.1 cm
37 in.
Class 10 (ISO 4)
Product Suggestions
Product Suggestions