missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ Tyvek™ IsoClean™ Series 105 White Coveralls
Ideal for use in controlled environments to protect against particulates and other contaminants.
Supplier: DuPont™ IC105SWHLG0025CS
Description
Light and durable coveralls provide coverage, including feet, while staying comfortable. Use in controlled environments with high standards for particle and microbiological contamination control.
- Clean processed and sterile
- Spinning process creates barrier to particles, micro-organisms, and non-hazardous liquids
- Lightweight, comfortable and durable
- Lowest level particle shedding
- Recyclable
- Serged seams
- Standard hood, elastic opening
- Elastic wrists and ankles
- Attached thumb loops
- Zipper closure
- Attached boots, PVC sole
Pharmaceutical, biotech, medical device, electronic settings
Specifications
| Cleanroom Coveralls | |
| Unisex | |
| White | |
| Thumb Loop | |
| Elastic | |
| Serged | |
| Elastic wrist and ankle, attached thumb loops, standard hood with elastic opening, attached boot with PVC soles, set sleeves | |
| 38 to 40 in. | |
| 96.52 to 101.6 cm | |
| 5 mil | |
| 35 in. | |
| 5.8 to 6 ft. |
| Large | |
| Tyvek | |
| Elastic, Attached Boots | |
| Hooded | |
| Zipper | |
| Sterile | |
| IC105SWH | |
| 29 in. | |
| 73.6 cm | |
| Barrier to dry particles, microorganisms, and non-hazardous liquids | |
| 88.9 cm | |
| 176.78 to 182.88 cm |