missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Tyvek™ IsoClean™ Series 105 White Coveralls
GREENER_CHOICE

Catalog No. p-4379412
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
Height (English):
5.11 to 6.3 ft.
5.5 to 5.10 ft.
5.8 to 6 ft.
6.1 to 6.5 ft.
6.2 to 6.6 ft.
Height (Metric):
155.75 to 192.04 cm
167.64 to 155.44 cm
176.78 to 182.88 cm
185.92 to 198.12 cm
188.97 to 201.16 cm
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Height (English) Height (Metric)
17685210 Medium 5.5 to 5.10 ft. 167.64 to 155.44 cm
17685211 Large 5.8 to 6 ft. 176.78 to 182.88 cm
17685212 X-Large 5.11 to 6.3 ft. 155.75 to 192.04 cm
17685213 2X-Large 6.1 to 6.5 ft. 185.92 to 198.12 cm
17685214 3X-Large 6.2 to 6.6 ft. 188.97 to 201.16 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 17685210 Supplier DuPont™ Supplier No. IC105SWHMD0025CS
Only null left

Ideal for use in controlled environments to protect against particulates and other contaminants.

Light and durable coveralls provide coverage, including feet, while staying comfortable. Use in controlled environments with high standards for particle and microbiological contamination control.

  • Clean processed and sterile
  • Spinning process creates barrier to particles, micro-organisms, and non-hazardous liquids
  • Lightweight, comfortable and durable
  • Lowest level particle shedding
  • Recyclable
  • Serged seams
  • Standard hood, elastic opening
  • Elastic wrists and ankles
  • Attached thumb loops
  • Zipper closure
  • Attached boots, PVC sole

Pharmaceutical, biotech, medical device, electronic settings

Specifications

Product Type Cleanroom Coveralls
Size Medium
Gender Unisex
Material Tyvek
Color White
Ankle Style Elastic, Attached Boots
Cuff Style Thumb Loop
Neck Style Hooded
Hood Type Elastic
Closure Type Zipper
Seam Type Serged
Sterility Sterile
Description Elastic wrist and ankle, attached thumb loops, standard hood with elastic opening, attached boot with PVC soles, set sleeves
Model IC105SWH
Size (English) Chest 36 to 38 in.
Size (English) Inseam 28 in.
Size (Metric) Chest 91.44 to 96.52 cm
Size (Metric) Inseam 71 cm
Thickness 5 mil
For Use With (Application) Barrier to dry particles, microorganisms, and non-hazardous liquids
Length (English) Sleeve 33.25 in.
Length (Metric) Sleeve 84.45 cm
Height (English) 5.5 to 5.10 ft.
Height (Metric) 167.64 to 155.44 cm
Show More Show Less