Learn More
DuPont™ Tyvek™ 400 Coverall (all)
Comfortable and durable flash spun high density polyethylene coveralls provides an inherent barrier against particles.
Supplier:  DuPont™ TY121SWH6X0025NS
 View more versions of this product
Catalog No. 17-500-195
Only null left
Explore available promotions
Alert:
Garments made of Tyvek should not be used around heat, flames, sparks, or in potentially flammable or explosive environments.
Description
Description
  • Pin locking slider locks in place when the pins on the puller are pushed into the zipper elements
  • Multiple interlocking threads are sewn around the raw edges of garment material to create a strong, stress-resistant seam
  • Laydown collar
  • Elastic opening for tighter fit at wrist
  • Attached Tyvek™ FC boots have special coating to provide added skid-resistance
  • Manufactured under specifications that do not contain natural rubber latex
Compliance
Meets USDA, FDA, OSHA standards; ASTM D1777, ASTM D3776, ASTM D774, ASTM D5733, ASTM D5034, ASTM D257, AATCC 127 compliant.
Specifications
Specifications
General Purpose Coveralls
Unisex
White
Elastic, Attached Boots
Collared
Front Zipper
Non-sterile
USDA, FDA, OSHA, ASTM D1777, ASTM D3776, ASTM D774, ASTM D5733, ASTM D5034, ASTM D257, AATCC 127
Lead and Asbestos Abatement, Remediation, General Maintenance, Spray Painting, Cleaning
6X-Large
Tyvek
Limited-use
Elastic
Unhooded
Serged
Laydown
TY121S
Product Suggestions
Product Suggestions