Learn More
DuPont™ Tychem™ 4000 Hooded Coverall
Supplier:  DuPont™ SL128TWHSM000600
 View more versions of this product
Catalog No. 19-162-786
Only null left
Explore available promotions
Description
Description
  • Zipper features a semi-automatic locking slider allowing the puller to stay down and in a locked position
  • Meets the U.S. industry requirements for blood (ASTM F1670) and viral penetration (ASTM F1671) protecting against several bloodborne pathogen exposure risks.
  • Taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing.
  • Attached respirator-fit hood with elastic around face opening, designed to cover neck and chin and fit around respirator face mask
  • Strong zipper for tough, flexible closure
  • Storm flap covers zipper which can be sealed by the wearer with adhesive strip to prevent intrusion at zipper
  • Elastic opening for tighter fit at wrist
  • Integrated socks composed of garment material
  • Attached flaps are designed to cover boot tops to help reduce potential for liquid intrusion
  • Manufactured under specifications that do not contain natural rubber latex
Specifications
Specifications
Small
Tychem
Attached Socks, Boot Overflaps
Attached
Taped
Unisex
White
Hooded
Front Zipper
Waste management, hazardous response and nuclear,environments
Product Suggestions
Product Suggestions