missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Tychem™ 4000 Hooded Coverall

Catalog No. p-4916677
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4X-Large
5X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19162787 X-Large
19162784 Large
19162785 Medium
19162786 Small
19162783 5X-Large
19162782 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 19162787 Supplier DuPont™ Supplier No. SL128TWHXL000600
Only null left
  • Zipper features a semi-automatic locking slider allowing the puller to stay down and in a locked position
  • Meets the U.S. industry requirements for blood (ASTM F1670) and viral penetration (ASTM F1671) protecting against several bloodborne pathogen exposure risks.
  • Taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing.
  • Attached respirator-fit hood with elastic around face opening, designed to cover neck and chin and fit around respirator face mask
  • Strong zipper for tough, flexible closure
  • Storm flap covers zipper which can be sealed by the wearer with adhesive strip to prevent intrusion at zipper
  • Elastic opening for tighter fit at wrist
  • Integrated socks composed of garment material
  • Attached flaps are designed to cover boot tops to help reduce potential for liquid intrusion
  • Manufactured under specifications that do not contain natural rubber latex

Specifications

Size X-Large
Gender Unisex
Material Tychem
Color White
Ankle Style Attached Socks, Boot Overflaps
Neck Style Hooded
Hood Type Attached
Closure Type Front Zipper
Seam Type Taped
For Use With (Application) Waste management, hazardous response and nuclear,environments