Learn More
DuPont™ Tychem™ 4000 Hooded Coverall
Supplier:  DuPont™ SL128TWH5X000600
 View more versions of this product
Catalog No. 19-162-783
Only null left
Explore available promotions
Description
Description
  • Zipper features a semi-automatic locking slider allowing the puller to stay down and in a locked position
  • Meets the U.S. industry requirements for blood (ASTM F1670) and viral penetration (ASTM F1671) protecting against several bloodborne pathogen exposure risks.
  • Taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing.
  • Attached respirator-fit hood with elastic around face opening, designed to cover neck and chin and fit around respirator face mask
  • Strong zipper for tough, flexible closure
  • Storm flap covers zipper which can be sealed by the wearer with adhesive strip to prevent intrusion at zipper
  • Elastic opening for tighter fit at wrist
  • Integrated socks composed of garment material
  • Attached flaps are designed to cover boot tops to help reduce potential for liquid intrusion
  • Manufactured under specifications that do not contain natural rubber latex
Specifications
Specifications
5X-Large
Tychem
Attached Socks, Boot Overflaps
Attached
Taped
Unisex
White
Hooded
Front Zipper
Waste management, hazardous response and nuclear,environments
Product Suggestions
Product Suggestions