Learn More
DuPont™ Tychem™ RESPONDER™ CSM Hooded Coverall
Coverall with respirator fit hood, attached butyl gloves and attached socks with outer boot flaps
Supplier:  DuPont™ RC128TTN2X000100
 View more versions of this product
Catalog No. 19-067-021
Only null left
Explore available promotions
Description
Description
  • Zipper features a semi-automatic locking slider allowing the puller to stay down and in a locked position
  • Made in the USA, North American Free Trade Agreement (NAFTA) compliant and Trade Agreement Act (TAA) compliant
  • Attached respirator-fit hood with elastic around face opening, designed to cover neck and chin and fit around respirator face mask
  • Strong zipper for tough, flexible closure
  • Longer zipper extends to chin for complete coverage of neck area
  • Storm flap covers zipper which can be secured by the wearer with rugged hook and loop material to prevent intrusion at zipper
  • MIL-G-12223 Butyl gloves provide mechanical and physical durability. Gloves are attached with vapor tight ring and clamp system and are replaceable
  • Integrated socks composed of garment material
  • Attached flaps are designed to cover boot tops to help reduce potential for liquid intrusion
  • Manufactured under specifications that do not contain natural rubber latex
Compliance
North American Free Trade Agreement (NAFTA) compliant and Trade Agreement Act (TAA) compliant
Specifications
Specifications
Tan
2X-Large
Zipper
6 ft. to 6 ft. 4 in.
28 in.
Tychem
Boot Overflaps
Unisex
Taped
Product Suggestions
Product Suggestions