missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ ProShield™ 60 Hooded Coverall
Coveralls with attached skid-resistant boots are suitable for protection against non-hazardous particles, light liquid splash and aerosols in many industrial applications.
$273.43 - $273.43
Specifications
| Product Type | General Purpose Coveralls |
|---|---|
| Gender | Unisex |
| Material | ProShield |
| Color | White |
| Disposable | Single-use |
| Catalog Number | Mfr. No. | Size | Height (English) | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Size | Height (English) | Price | Quantity | ||||
|
29-634-236
|
DuPont™
NG122SWH2X002500 |
2X-Large | 6 to 6.4 ft. |
Case of 25 Each for $273.43
|
|
||||
Description
- Pin locking slider locks in place when the pins on the puller are pushed into the zipper elements
- Attached hood with elastic around face opening.
- Front zipper closure for easy donning and doffing.
- Elastic opening for tighter fit at wrist
- Attached ProShield™ 3 boots have special coating to provide added skid-resistance
- Manufactured under specifications that do not contain natural rubber latex
Compliance
Meets USDA, FDA, OSHA standards; ASTM D1777, ASTM D3776, ASTM D774, ASTM D5733, ASTM D5034, ASTM D257, AATCC 127 compliant.
- Measured per ASTM™ F 903 in relation to the following substances: household bleach, diazinon (4 oz./gal.), hydraulic jack oil, machine oil, malathion (50%), motor oil (40W), sodium hydroxide and isocyanate-based paint hardener. Tests were performed by independent laboratories on material samples under laboratory conditions, not with actual garments or in actual onsite conditions.
- ASTM D 751 (Grab Tensile Strength), ASTM D 1117 (Trapezoid Tear), ASTM D 3787 (Ball Burst).
- Certified incinerable by independent laboratory. For details, contact your Fisher Safety Customer Service Representative at 1-800-772-6733.
Specifications
| General Purpose Coveralls | |
| ProShield | |
| Single-use | |
| Elastic | |
| Standard, Elastic Face | |
| Serged | |
| Particle, Liquid | |
| Microporous Film, Laminated to a Spundbonded Polypropylene Substrate | |
| NG120SWH | |
| 25/Cs. |
| Unisex | |
| White | |
| Elastic, Attached Boots | |
| Hooded | |
| Front Zipper | |
| Non-sterile | |
| USDA, FDA, OSHA, ASTM D1777, ASTM D3776, ASTM D774, ASTM D5733, ASTM D5034, ASTM D257, AATCC 127 | |
| Yes | |
| Automotive Refinishing, Waste Cleanup, Asbestos Abatement, Hazardous Remediation |