missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ ProShield™ 60 Hooded Coverall

Catalog No. 29634236
Change view
Click to view available options
Size:
2X-Large
Height (English):
6 to 6.4 ft.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Height (English)
29634236 2X-Large 6 to 6.4 ft.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 29634236 Supplier DuPont™ Supplier No. NG122SWH2X002500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Coveralls with attached skid-resistant boots are suitable for protection against non-hazardous particles, light liquid splash and aerosols in many industrial applications.

  • Pin locking slider locks in place when the pins on the puller are pushed into the zipper elements
  • Attached hood with elastic around face opening.
  • Front zipper closure for easy donning and doffing.
  • Elastic opening for tighter fit at wrist
  • Attached ProShield™ 3 boots have special coating to provide added skid-resistance
  • Manufactured under specifications that do not contain natural rubber latex

Compliance

Meets USDA, FDA, OSHA standards; ASTM D1777, ASTM D3776, ASTM D774, ASTM D5733, ASTM D5034, ASTM D257, AATCC 127 compliant.

  1. Measured per ASTM™ F 903 in relation to the following substances: household bleach, diazinon (4 oz./gal.), hydraulic jack oil, machine oil, malathion (50%), motor oil (40W), sodium hydroxide and isocyanate-based paint hardener. Tests were performed by independent laboratories on material samples under laboratory conditions, not with actual garments or in actual onsite conditions.
  2. ASTM D 751 (Grab Tensile Strength), ASTM D 1117 (Trapezoid Tear), ASTM D 3787 (Ball Burst).
  3. Certified incinerable by independent laboratory. For details, contact your Fisher Safety Customer Service Representative at 1-800-772-6733.

Specifications

Product Type General Purpose Coveralls
Size 2X-Large
Gender Unisex
Material ProShield
Color White
Disposable Single-use
Waist Style Non-elastic
Ankle Style Elastic, Attached Boots
Cuff Style Elastic
Neck Style Hooded
Hood Type Standard, Elastic Face
Closure Type Front Zipper
Seam Type Serged
Sterility Non-sterile
Resistance Particle, Liquid
Certifications/Compliance USDA, FDA, OSHA, ASTM D1777, ASTM D3776, ASTM D774, ASTM D5733, ASTM D5034, ASTM D257, AATCC 127
Description Microporous Film, Laminated to a Spundbonded Polypropylene Substrate
Latex Free Yes
Model NG120SWH
Size (English) Chest 44.25 to 47.75 in.
Size (English) Inseam 28.25 in.
Size (Metric) Chest 112.39 to 121.28 cm
Size (Metric) Inseam 71.75 cm
Thickness 10 mil
For Use With (Application) Automotive Refinishing, Waste Cleanup, Asbestos Abatement, Hazardous Remediation
Length (English) Sleeve 35.75 in.
Length (Metric) Sleeve 90.8 cm
Height (English) 6 to 6.4 ft.
Height (Metric) 1.82 to 1.95 m
Quantity 25/Cs.
Show More Show Less