missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ Tyvek™ IsoClean™ Frock
Clean-processed and sterile frocks provide an inherent barrier to particles, microorganisms, and non-hazardous light liquid splash.
Supplier: DuPont™ IC270BWH3X0030CS
Alert:
Garments made of Tyvek should not be used around heat, flames, sparks or in potentially flammable or explosive environments.
Description
- Garments are gamma sterilized to SAL of 10-6.
- Clean-processed garments offer lowest level of particle shedding within DuPont product portfolio
- Bound seams are covered with garment fabric to reinforce the seam and to reduce the potential for particle penetration
- Elastic opening for tighter fit at wrist
- Bound neck for lower particle shedding
- Adjustable snap at neck for better fit
- Full traceability on all sterilized apparel with Certificates of Sterility.
Specifications
| Controlled Environments Frock | |
| Tyvek | |
| Single-use | |
| Snaps | |
| Bound | |
| IsoClean™ 270 Series | |
| 109 to 116 cm | |
| Clean-processed, Individually Packaged | |
| 114.3 cm | |
| 100.96 cm |
| 3X-Large | |
| White | |
| Sterile | |
| Elastic | |
| None | |
| 43 to 46 in. | |
| Long Sleeve | |
| 45 in. | |
| 39.75 in. | |
| Class 100 (ISO 5) |