missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DUOX2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5150265
Description
DUOX2 Polyclonal Antibody for Western Blot, ICC/IF, IHC
The protein encoded by this gene is a glycoprotein and a member of the NADPH oxidase family. The synthesis of thyroid hormone is catalyzed by a protein complex located at the apical membrane of thyroid follicular cells. This complex contains an iodide transporter, thyroperoxidase, and a peroxide generating system that includes this encoded protein and DUOX1. This protein is known as dual oxidase because it has both a peroxidase homology domain and a gp91phox domain.Specifications
| DUOX2 | |
| Polyclonal | |
| Unconjugated | |
| DUOX2 | |
| Dual oxidase 2; dual oxidase-like domains 2; DUOX2; flavoprotein NADPH oxidase; large NOX 2; LNOX2; Long NOX 2; NADH/NADPH thyroid oxidase p138-tox; NADPH oxidase/peroxidase DUOX2; NADPH thyroid oxidase 2; nicotinamide adenine dinucleotide phosphate oxidase; NOXEF2; p138 thyroid oxidase; P138-TOX; TDH6; THOX2; Thyroid oxidase 2 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 214593, 79107 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q9ES45 | |
| DUOX2 | |
| Recombinant Dual Oxidase 2 (DUOX2) (Asp1229-Phe1517). | |
| 200 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody |