missing translation for 'onlineSavingsMsg'
Learn More

Duloxetine Hydrochloride Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111013007
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013007 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013007 Supplier Merck Emd Millipore Supplier No. PHR1865500MG

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Specifications

CAS 136434-34-9
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C18H19 NOS · HCl
MDL Number MFCD06407958
Synonym -Duloxetine hydrochloride; (gammaS)-N-Methyl-gamma-(1-naphthalenyloxy)-2-thiophenepropanamine hydrochloride; (+)--N-Methyl-gamma-(1-naphthyloxy)-2-thiophenepropylamine hydrochloride; (+)-N-Methyl-3-(1-naphthalenyloxy)-3-(2-thienyl)propanamine hydrochloride; LY-248686 hydrochloride
Type Certified Reference Material
Quantity 500 mg
Formula Weight 333.88
Grade Certified Reference Material