missing translation for 'onlineSavingsMsg'
Learn More
Learn More
dsRNA Mouse anti-Virus, Unconjugated, Clone: rJ2, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABE1134100UL
Description
Anti-dsRNA, clone rJ2, Cat. No. MABE1134, is a mouse monoclonal antibody that detects double stranded RNA (dsRNA) and has been tested for use in Immunocytochemistry and Immunofluorescence.Specifications
| dsRNA | |
| Monoclonal | |
| Please refer to lot specific datasheet. | |
| Mouse monoclonal antibody in supernatant without preservatives. | |
| N/A | |
| Double stranded RNA produced by positive sense genome viruses. | |
| Unpurified | |
| RUO | |
| Primary | |
| Virus | |
| Antibodies | |
| IgG2a κ |
| Immunocytochemistry, Immunofluorescence | |
| rJ2 | |
| Unconjugated | |
| Double-stranded RNA | |
| Mouse | |
| N/A | |
| 100 μL | |
| Inflammation & Immunology | |
| Clone rJ2 specifically recognizes double stranded RNA (dsRNA) of greater than 40 bp in length that is generated during the replication of positive sense genome viruses. | |
| Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance. | |
| Culture Supernatant |