missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DSC1 Recombinant Rabbit Monoclonal Antibody (23GB5210)

Catalog No. MA553865
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA553865 20 μL
MA553864 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA553865 Supplier Invitrogen™ Supplier No. MA553865
Only null left

Rabbit Recombinant Monoclonal Antibody

DSC1 Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot

The desmosomal cadherin desmocollin DSC1 is expressed in upper epidermis where strong adhesion is required. DSC1 is a type I membrane protein (4) required for strong adhesion and barrier maintenance in epidermis and contributes to epidermal differentiation.The DSC1 gene comprises 17 exons spanning approximately 33 kb on 18q12.1.

Specifications

Antigen DSC1
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB5210
Concentration 1.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene DSC1
Gene Accession No. P55849, Q08554
Gene Alias 1110020A10Rik; AI507491; Cadherin family member 1; CDHF1; desmocollin 1; Desmocollin1; desmocollin-1; Desmosomal glycoprotein 2/3; DG2/DG3; DSC1; Dsc1a; Dsc1b
Gene Symbols DSC1
Host Species Rabbit
Immunogen A synthesized peptide derived from human Desmocollin 1 (824-894AA).
Purification Method Affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13505, 1823, 291759
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less