missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ DS-33 Matrix Standard Set for the 310 Genetic Analyzer

Catalog No. 4318159
Encompass
Change view
Click to view available options
For Use With (Equipment):
310 Genetic Analyzer
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
4318159 310 Genetic Analyzer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 4318159 Supplier Applied Biosystems™ Supplier No. 4318159

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Contains 5 Dye Labeled Oligonucleotides used for Generating the Multicomponent Matrix in a 1X TE Solution.

The dye-labeled oligonucleotides included in the matrix standard set DS-33 [6-FAM™, VIC™, NED™, PET™, and LIZ™] are used to generate the 'multi-component matrix' required for the ABI PRISM™ 310 genetic analyzer under denaturing applications.

  • Matrix standards do not need to be run with every set of sample injections
  • This set of five matrix standards only need to be run once to generate a matrix file which is then applied to samples run under similar conditions

Order Info

10 Runs

Specifications

Content And Storage Contains 5 dye labeled oligonucleotides used for generating the multicomponent matrix in a 1X TE solution. Store the kit at 2 - 8°C. Do not freeze.
For Use With (Equipment) 310 Genetic Analyzer
For Use With (Application) Sequencing
Label or Dye FAM, LIZ, NED, PET, VIC
No. of Reactions 10 Runs
Product Type DS-33 Matrix Standard Set
Shipping Condition Wet Ice

For research use only